Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: C17orf75 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_022344.3 | NP_071739.2 | 6 | 678 | ||
XM_005258022.3 | XP_005258079.1 | 7 | 696 | |||
XM_017024940.1 | XP_016880429.1 | 6 | 547 | |||
XM_017024941.1 | XP_016880430.1 | 6 | 545 | |||
GenBank mRNA | AF052093.1 | - | 6 | 605 | ||
AF305686.1 | - | 6 | 634 | |||
BC026017.2 | - | 6 | 624 | 3352761 | ||
BC035715.1 | - | 6 | 632 | 5588164 | ||
BC047325.1 | - | 6 | 618 | 5493477 | ||
BC054035.1 | - | 6 | 612 | 6143106 |