Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: STRA6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001142617.1 | NP_001136089.1 | 18 | 1948 | ||
NM_001142618.1 | NP_001136090.1 | 18 | 1930 | |||
NM_001142619.1 | NP_001136091.1 | 18 | 1921 | |||
NM_001199040.1 | NP_001185969.1 | 18 | 2064 | |||
NM_001199041.1 | NP_001185970.1 | 18 | 2075 | |||
NM_001199042.1 | NP_001185971.1 | 18 | 2179 | |||
NM_022369.3 | NP_071764.3 | 18 | 1884 | |||
XM_011521883.1 | XP_011520185.1 | 18 | 1850 | |||
XM_011521884.1 | XP_011520186.1 | 18 | 1991 | |||
XM_017022478.1 | XP_016877967.1 | 18 | 1799 | |||
XM_017022479.1 | XP_016877968.1 | 18 | 1890 | |||
XM_017022480.1 | XP_016877969.1 | 18 | 1914 | |||
GenBank mRNA | AF352728.1 | - | 18 | 1750 | ||
AF352729.1 | - | 18 | 1860 | |||
AF370419.2 | - | 18 | 1856 | |||
AK022603.1 | - | 14 | 1434 | |||
AK056125.1 | - | 18 | 1945 | |||
AK091152.1 | - | 18 | 1974 | |||
AK092227.1 | - | 13 | 1411 | |||
AK291966.1 | - | 18 | 1820 | |||
AK298921.1 | - | 18 | 1819 | |||
AK298928.1 | - | 16 | 1488 | |||
AK299191.1 | - | 18 | 2064 | |||
AK302932.1 | - | 18 | 2075 | |||
AK309676.1 | - | 18 | 2159 | |||
AY358748.1 | - | 18 | 1860 | |||
AY359089.1 | - | 18 | 1750 | |||
BC025256.1 | - | 18 | 1856 | 4892821 | ||
BX537413.1 | - | 18 | 1826 |