Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: C16orf58 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_022744.3 | NP_073581.2 | 5 | 573 | ||
GenBank mRNA | AK023930.1 | - | 5 | 539 | ||
AK222913.1 | - | 5 | 539 | |||
AK296166.1 | - | 4 | 423 | |||
AK304608.1 | - | 6 | 401 | |||
BC009308.2 | - | 5 | 533 | 4301507 |