Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: C16orf58 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_022744.3 | NP_073581.2 | 12 | 1317 | ||
XM_017023570.1 | XP_016879059.1 | 10 | 1148 | |||
GenBank mRNA | AK023930.1 | - | 12 | 1283 | ||
AK092580.1 | - | 3 | 945 | |||
AK222913.1 | - | 12 | 1283 | |||
AK304608.1 | - | 13 | 1145 | |||
BC000822.1 | - | 6 | 560 | |||
BC009308.2 | - | 12 | 1277 | 4301507 |