Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: C16orf58 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_022744.3 | NP_073581.2 | 5, 6 | 645 | ||
GenBank mRNA | AK000679.1 | - | 1, 2 | 66 | ||
AK023930.1 | - | 5, 6 | 611 | |||
AK222913.1 | - | 5, 6 | 611 | |||
AK296166.1 | - | 4, 5 | 495 | |||
AK304608.1 | - | 6, 7 | 473 | |||
BC009308.2 | - | 5, 6 | 605 | 4301507 |