Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CHN2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001293069.1 | NP_001279998.1 | 6 | 501 | ||
NM_001293070.1 | NP_001279999.1 | 6 | 715 | |||
NM_001293071.1 | NP_001280000.1 | 4 | 620 | |||
NM_001293072.1 | NP_001280001.1 | 5 | 433 | |||
NM_004067.3 | NP_004058.1 | 5 | 676 | |||
XM_011515105.2 | XP_011513407.1 | 8 | 556 | |||
XM_011515106.2 | XP_011513408.1 | 7 | 517 | |||
XM_011515107.2 | XP_011513409.1 | 7 | 1299 | |||
XM_017011721.1 | XP_016867210.1 | 9 | 574 | |||
XM_017011722.1 | XP_016867211.1 | 8 | 1317 | |||
GenBank mRNA | AK124021.1 | - | 1 | 1784 | ||
AK293959.1 | - | 6 | 295 | |||
AK294032.1 | - | 5 | 433 | |||
AK294271.1 | - | 6 | 502 | |||
AK313021.1 | - | 5 | 450 | |||
BC112155.1 | - | 5 | 421 | 8327623 | ||
DA190643.1 | - | 6 | 501 | |||
EU732760.1 | - | 4 | 183 | |||
EU732764.1 | - | 5 | 239 | |||
EU732765.1 | - | 5 | 239 | |||
EU732766.1 | - | 5 | 239 | |||
GQ924106.1 | - | 6 | 502 | |||
U28926.1 | - | 5 | 473 |