Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FRY | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_023037.2 | NP_075463.2 | 2 | 742 | ||
XM_006719749.3 | XP_006719812.1 | 2 | 1823 | |||
XM_017020303.1 | XP_016875792.1 | 2 | 2300 | |||
XM_017020304.1 | XP_016875793.1 | 2 | 742 | |||
XM_017020305.1 | XP_016875794.1 | 2 | 2319 | |||
GenBank mRNA | AL049784.1 | - | 2 | 742 | ||
U50534.1 | - | 2 | 741 |