Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PRR14 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001320464.1 | NP_001307393.1 | 10 | 1531 | ||
NM_024031.3 | NP_076936.1 | 10 | 1687 | |||
GenBank mRNA | AK074783.1 | - | 10 | 1542 | ||
AK125716.1 | - | 8 | 1634 | |||
BC000119.1 | - | 10 | 1554 | 3351963 | ||
BC010232.1 | - | 8 | 1296 | |||
BC021934.1 | - | 9 | 1639 | 4876482 | ||
BC050677.1 | - | 10 | 1634 | 6053861 | ||
CR936781.1 | - | 8 | 1145 |