Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CS | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_004077.2 | NP_004068.2 | 3 | 327 | ||
GenBank mRNA | AF047042.1 | - | 3 | 137 | ||
AF053631.1 | - | 3 | 302 | |||
AF147410.1 | - | 1 | 27 | |||
AK074956.1 | - | 3 | 175 | |||
AK293263.1 | - | 1 | 112 | |||
AK296249.1 | - | 4 | 307 | |||
AL713477.1 | - | 3 | 324 | |||
BC010106.1 | - | 3 | 255 | 3542491 | ||
BC072016.1 | - | 3 | 170 | 6484456 | ||
BI258893.1 | - | 4 | 354 | |||
CB990178.1 | - | 3 | 354 |