Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DHX58 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_024119.2 | NP_077024.2 | 14 | 2548 | ||
XM_017025059.1 | XP_016880548.1 | 13 | 2481 | |||
XM_017025060.1 | XP_016880549.1 | 14 | 2544 | |||
GenBank mRNA | AK021416.1 | - | 14 | 2548 | ||
AK097669.1 | - | 12 | 2189 | |||
AK225549.1 | - | 12 | 1944 | |||
BC014949.1 | - | 14 | 2485 | 4865798 | ||
BM723672.1 | - | 2 | 543 |