Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CORO7-PAM16 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001201479.1 | NP_001188408.1 | 4, 5 | 753 | ||
GenBank mRNA | AL833954.1 | - | 5, 6 | 776 |
Gene Symbol: CORO7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001201472.1 | NP_001188401.1 | 4, 5 | 699 | ||
NM_024535.4 | NP_078811.3 | 4, 5 | 753 | |||
GenBank mRNA | AK025674.1 | - | 4, 5 | 362 | ||
AK094596.1 | - | 4, 5 | 433 | |||
AK294045.1 | - | 4, 5 | 334 | |||
AK296723.1 | - | 4, 5 | 331 | |||
AK309327.1 | - | 4, 5 | 388 | |||
AK309763.1 | - | 2, 3 | 172 | |||
BC117289.1 | - | 4, 5 | 372 | 40125840 | ||
BC117291.1 | - | 4, 5 | 372 | 40125842 | ||
DA094935.1 | - | 4, 5 | 430 |