Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CARS2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_024537.3 | NP_078813.1 | 3, 4 | 480 | ||
XM_006719953.2 | XP_006720016.1 | 3, 4 | 560 | |||
XM_011521117.2 | XP_011519419.1 | 3, 4 | 416 | |||
XM_011521118.2 | XP_011519420.1 | 3, 4 | 416 | |||
XM_017020741.1 | XP_016876230.1 | 2, 3 | 208 | |||
XM_017020742.1 | XP_016876231.1 | 3, 4 | 416 | |||
GenBank mRNA | AB062436.1 | - | 3, 4 | 391 | ||
AK022180.1 | - | 3, 4 | 416 | |||
AK301907.1 | - | 2, 3 | 585 | |||
BC007220.1 | - | 3, 4 | 403 | 2822557 | ||
DA966145.1 | - | 3, 4 | 435 | |||
HY160154.1 | - | 3, 4 | 462 |