Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CDMO-CRO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other TCTN1 SIRNA Assays ›
Gene Symbol:
TCTN1
Gene Name:
tectonic family member 1
Gene Aliases:
JBTS13, TECT1
Chromosome Location:
Chr.12: 110614027 - 110649130 on Build GRCh38
UniGene ID:
Hs.211511
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
140812
Purification
HPLC
Standard
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: TCTN1
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001082537.2
NP_001076006.1
3
596
NM_001082538.2
NP_001076007.1
3
596
NM_001173975.2
NP_001167446.1
3
657
NM_001173976.1
NP_001167447.1
4
672
NM_001319680.1
NP_001306609.1
3
596
NM_001319681.1
NP_001306610.1
3
596
NM_001319682.1
NP_001306611.1
3
657
NM_024549.5
NP_078825.2
3
596
NR_135088.1
3
657
XM_005253934.3
XP_005253991.1
3
500
XM_005253935.3
XP_005253992.1
3
500
XM_006719594.2
XP_006719657.1
3
561
XM_006719595.2
XP_006719658.1
4
667
XM_006719596.2
XP_006719659.1
3
517
XM_006719597.3
XP_006719660.1
2
387
XM_006719598.2
XP_006719661.1
3
517
XM_006719599.2
XP_006719662.1
3
495
XM_006719600.2
XP_006719663.1
2
434
XM_011538733.2
XP_011537035.1
3
500
XM_011538734.2
XP_011537036.1
2
455
XM_011538735.1
XP_011537037.1
3
500
XM_011538737.2
XP_011537039.1
3
500
XM_011538738.2
XP_011537040.1
3
500
XM_017019964.1
XP_016875453.1
3
561
XM_017019965.1
XP_016875454.1
3
484
XM_017019966.1
XP_016875455.1
3
531
XM_017019967.1
XP_016875456.1
3
361
XM_017019968.1
XP_016875457.1
2
450
XM_017019969.1
XP_016875458.1
3
495
GenBank mRNA
AK024780.1
-
4
570
AK055891.1
-
3
510
AK092775.1
-
3
558
AK295514.1
-
3
596
AK301732.1
-
3
461
AY358184.1
-
3
459
BC033811.1
-
3
543
BC040113.1
-
3
486
6054789
BC044885.1
-
3
519
BC062611.1
-
3
447
5184202
DN997186.1
-
3
556
DQ278868.1
-
3
521
Back To Top
More Information
Gene Ontology Categories:
Function(s)
in utero embryonic development
neural tube formation
regulation of smoothened signaling pathway
somatic motor neuron differentiation
telencephalon development
dorsal/ventral neural tube patterning
central nervous system interneuron axonogenesis
cilium morphogenesis
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-blue-5c9c5c4749-jlg4l:80/100.66.77.222:80.
git-commit: d22414ba47b22cc02ffc24003f0e179da61511a6
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.21.0-Offline