Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: EFL1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001322845.1 | NP_001309774.1 | 3, 4 | 573 | ||
NM_024580.5 | NP_078856.4 | 3, 4 | 331 | |||
NR_136410.1 | 3, 4 | 331 | ||||
GenBank mRNA | AK056656.1 | - | 3, 4 | 261 | ||
AK300348.1 | - | 3, 4 | 241 |
Gene Symbol: EFTUD1P1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_036652.1 | 3, 4 | 311 | |||
GenBank mRNA | DB064528.1 | - | 3, 4 | 311 |