Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: AAGAB | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001271885.1 | NP_001258814.1 | 10 | 2707 | ||
NM_001271886.1 | NP_001258815.1 | 10 | 2968 | |||
NM_024666.4 | NP_078942.3 | 10 | 2823 | |||
XM_011522020.1 | XP_011520322.1 | 10 | 3073 | |||
GenBank mRNA | AL136715.1 | - | 10 | 2733 | ||
BQ185328.1 | - | 1 | 252 | |||
CB243401.1 | - | 1 | 253 |