Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other SLTM SIRNA Assays ›
Gene Symbol:
SLTM
Gene Name:
SAFB like transcription modulator
Gene Aliases:
Met
Chromosome Location:
Chr.15: 58879045 - 58934313 on Build GRCh38
UniGene ID:
Hs.512932
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
140879
Purification
HPLC
Standard
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: SLTM
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001013843.2
NP_001013865.1
15
2004
NM_024755.3
NP_079031.2
15
2058
NR_135042.1
13
1391
NR_135043.1
13
1501
XM_006720686.3
XP_006720749.3
13
2449
XM_006720690.1
XP_006720753.1
11
1155
XM_011522022.1
XP_011520324.1
15
2716
XM_011522023.1
XP_011520325.1
15
2662
XM_011522024.1
XP_011520326.1
14
2566
XM_011522025.1
XP_011520327.1
14
2518
XM_011522026.2
XP_011520328.2
14
2497
XM_011522027.1
XP_011520329.1
13
2470
XM_011522028.1
XP_011520330.1
13
2416
XM_011522029.2
XP_011520331.1
15
1897
XM_011522030.2
XP_011520332.1
15
2716
XM_011522031.2
XP_011520333.1
12
1552
XM_017022576.1
XP_016878065.1
14
1854
XM_017022577.1
XP_016878066.1
14
2443
XM_017022578.1
XP_016878067.1
13
2395
XM_017022579.1
XP_016878068.1
15
2061
XM_017022580.1
XP_016878069.1
14
1515
GenBank mRNA
AK023275.1
-
5
512
AK055195.1
-
13
1294
AK291423.1
-
13
1424
BC014944.1
-
8
861
BC046119.1
-
4
353
BC140851.1
-
15
2025
9021719
Back To Top
More Information
Other Species Targeted
This siRNA also targets the following transcripts in Mouse: NM_025690, NM_026337, BC139202, CD806976, CN723089, and others.
Gene Ontology Categories:
Function(s)
Process(es)
transcription, DNA-templated
regulation of transcription, DNA-templated
apoptotic process
nucleotide binding
poly(A) RNA binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-blue-65d5d77f49-g5d4h:80/100.66.76.71:80.
git-commit: a4c025c8ba47546866237b65de1f276a5fdbbe78
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.18.1-Offline