Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: BORA | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001286746.1 | NP_001273675.1 | 9 | 1174 | ||
NM_001286747.1 | NP_001273676.1 | 8 | 969 | |||
NM_024808.3 | NP_079084.3 | 9 | 1076 | |||
XM_006719868.3 | XP_006719931.2 | 7 | 933 | |||
GenBank mRNA | AK026277.1 | - | 4 | 401 | ||
AK291496.1 | - | 9 | 902 | |||
AK298608.1 | - | 8 | 795 | |||
AK300758.1 | - | 8 | 874 | |||
BC025367.2 | - | 9 | 887 | 4098541 | ||
BC044213.1 | - | 8 | 700 | |||
BC056876.1 | - | 9 | 899 | 6473892 | ||
BX648605.1 | - | 9 | 1174 | |||
EU834129.1 | - | 9 | 982 |