Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SETD6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001160305.2 | NP_001153777.1 | 7 | 1055 | ||
NM_024860.3 | NP_079136.2 | 8 | 983 | |||
NR_134583.1 | 8 | 1014 | ||||
GenBank mRNA | AK024801.1 | - | 8 | 939 | ||
AK127136.1 | - | 4 | 2460 | |||
AK290495.1 | - | 7 | 1017 | |||
BC022451.1 | - | 7 | 1017 | 4794299 | ||
BC110433.1 | - | 8 | 998 |