Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ESRP2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_024939.2 | NP_079215.2 | 10, 11 | 1495 | ||
XM_005256153.4 | XP_005256210.1 | 10, 11 | 1434 | |||
XM_006721273.3 | XP_006721336.1 | 10, 11 | 1503 | |||
XM_011523338.2 | XP_011521640.1 | 9, 10 | 1395 | |||
GenBank mRNA | AF289577.1 | - | 4, 5 | 597 | ||
AK025571.1 | - | 10, 11 | 1424 | |||
BC030146.2 | - | 10, 11 | 1317 | 4810948 | ||
BC069241.1 | - | 10, 11 | 1846 | |||
LN680438.1 | - | 10, 11 | 1307 | |||
LN680439.1 | - | 10, 11 | 1307 |