Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NAA25 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_024953.3 | NP_079229.2 | 3, 4 | 345 | ||
XM_006719606.2 | XP_006719669.1 | 3, 4 | 1005 | |||
XM_017019977.1 | XP_016875466.1 | 1, 2 | 68 | |||
GenBank mRNA | AB054990.1 | - | 3, 4 | 287 | ||
AK292485.1 | - | 3, 4 | 318 | |||
AK307494.1 | - | 4, 5 | 494 | |||
AV760516.2 | - | 3, 4 | 310 | |||
BC034357.1 | - | 3, 4 | 299 | |||
BC065481.1 | - | 3, 4 | 278 | |||
BC113585.1 | - | 3, 4 | 345 | 8322637 | ||
BC113587.1 | - | 3, 4 | 345 | 8322639 | ||
BX641140.1 | - | 3, 4 | 300 | |||
BX647450.1 | - | 3, 4 | 298 |