Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CCDC92 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001304957.1 | NP_001291886.1 | 5 | 844 | ||
NM_001304958.1 | NP_001291887.1 | 6 | 951 | |||
NM_001304959.1 | NP_001291888.1 | 5 | 593 | |||
NM_001304960.1 | NP_001291889.1 | 7 | 762 | |||
NM_001304961.1 | NP_001291890.1 | 4 | 761 | |||
NM_025140.2 | NP_079416.1 | 5 | 854 | |||
XM_005253624.2 | XP_005253681.1 | 5 | 598 | |||
XM_017019984.1 | XP_016875473.1 | 5 | 1412 | |||
GenBank mRNA | AB015292.1 | - | 4 | 374 | ||
AK026124.1 | - | 5 | 485 | |||
AK124358.1 | - | 1 | 1943 | |||
AK125866.1 | - | 3 | 2372 | |||
AK222661.1 | - | 5 | 442 | |||
AW953473.1 | - | 8 | 733 | |||
BC017186.2 | - | 5 | 477 | 3051346 | ||
BX394686.2 | - | 4 | 510 | |||
DA370549.1 | - | 6 | 549 |