Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ACSF2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001288968.1 | NP_001275897.1 | 17 | 2093 | ||
NM_001288969.1 | NP_001275898.1 | 16 | 1979 | |||
NM_001288970.1 | NP_001275899.1 | 15 | 1889 | |||
NM_001288971.1 | NP_001275900.1 | 14 | 1693 | |||
NM_001288972.1 | NP_001275901.1 | 16 | 1935 | |||
NM_025149.5 | NP_079425.3 | 16 | 2018 | |||
NR_110232.1 | 6 | 651 | ||||
XM_011525294.1 | XP_011523596.1 | 16 | 1940 | |||
GenBank mRNA | AK024573.1 | - | 16 | 1925 | ||
AK225044.1 | - | 16 | 1925 | |||
AK225561.1 | - | 16 | 1924 | |||
AK294205.1 | - | 17 | 2035 | |||
AK295258.1 | - | 16 | 1887 | |||
AK300145.1 | - | 15 | 1797 | |||
AK300625.1 | - | 14 | 1693 | |||
AK308584.1 | - | 6 | 637 | |||
AY358660.1 | - | 16 | 1954 | |||
BC012053.1 | - | 16 | 1931 | 4577089 | ||
BC014123.2 | - | 16 | 1938 | 4560029 |