Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CCDC68 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001143829.1 | NP_001137301.1 | 12 | 1614 | ||
NM_025214.2 | NP_079490.1 | 12 | 1717 | |||
XM_006722552.3 | XP_006722615.1 | 12 | 1666 | |||
XM_011526198.2 | XP_011524500.1 | 12 | 2726 | |||
XM_011526199.2 | XP_011524501.1 | 12 | 1625 | |||
XM_011526200.2 | XP_011524502.1 | 12 | 1778 | |||
XM_011526201.2 | XP_011524503.1 | 12 | 2685 | |||
XM_011526202.1 | XP_011524504.1 | 11 | 1640 | |||
XM_011526203.2 | XP_011524505.1 | 11 | 1552 | |||
XM_017026011.1 | XP_016881500.1 | 11 | 1718 | |||
GenBank mRNA | AF273051.1 | - | 12 | 1614 |