Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PTGES3L | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001142653.1 | NP_001136125.1 | 5 | 796 | ||
NM_001142654.1 | NP_001136126.1 | 5 | 781 | |||
NM_001261430.1 | NP_001248359.1 | 6 | 895 | |||
GenBank mRNA | AK056965.1 | - | 5 | 337 | ||
AK093402.1 | - | 4 | 1205 | |||
BG776211.1 | - | 6 | 442 | |||
DA900052.1 | - | 5 | 349 |
Gene Symbol: PTGES3L-AARSD1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001136042.2 | NP_001129514.2 | 6 | 895 | ||
NM_025267.3 | NP_079543.1 | 6 | 511 | |||
GenBank mRNA | AF039235.1 | - | 6 | 600 | ||
AK094084.1 | - | 6 | 449 | |||
AK295447.1 | - | 6 | 895 | |||
BC004172.2 | - | 6 | 511 | 2823004 | ||
BC019324.1 | - | 6 | 511 | 2823004 |