Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CDT1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_030928.3 | NP_112190.2 | 7 | 1070 | ||
GenBank mRNA | AB053172.1 | - | 7 | 1070 | ||
AF070552.1 | - | 5 | 609 | |||
AF321125.1 | - | 7 | 1041 | |||
BC000137.2 | - | 7 | 1032 | 3048353 | ||
BC008676.1 | - | 7 | 908 | |||
BC008860.2 | - | 7 | 963 | |||
BC009410.1 | - | 7 | 1026 | 3613566 | ||
BC014202.2 | - | 3 | 331 | |||
BC021126.1 | - | 5 | 602 | |||
BC049205.1 | - | 5 | 674 |