Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GSG2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_031965.2 | NP_114171.2 | 1 | 1669 | ||
GenBank mRNA | AB039834.3 | - | 1 | 1666 | ||
AF289865.1 | - | 1 | 1665 | |||
AK292715.1 | - | 1 | 1655 | |||
AK310546.1 | - | 1 | 1669 | |||
AK315247.1 | - | 1 | 1669 | |||
BC016626.1 | - | 1 | 1655 | |||
BC047457.1 | - | 1 | 1650 | 5298590 |