Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TMEM117 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001286211.1 | NP_001273140.1 | 2 | 576 | ||
NM_001286212.1 | NP_001273141.1 | 2 | 576 | |||
NM_001286213.1 | NP_001273142.1 | 2 | 576 | |||
NM_032256.2 | NP_115632.1 | 2 | 576 | |||
XM_011538831.2 | XP_011537133.1 | 2 | 585 | |||
XM_011538832.2 | XP_011537134.1 | 2 | 260 | |||
XM_011538833.2 | XP_011537135.1 | 1 | 15 | |||
XM_017020023.1 | XP_016875512.1 | 2 | 302 | |||
GenBank mRNA | AK057287.1 | - | 2 | 295 | ||
AL136855.1 | - | 2 | 278 | |||
BC030284.1 | - | 2 | 284 | 5240522 | ||
BC060798.1 | - | 2 | 339 | 30348706 | ||
DA007410.1 | - | 1 | 15 |