Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TMEM107 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_032354.3 | NP_115730.2 | 5 | 578 | ||
NM_183065.2 | NP_898888.1 | 5 | 560 | |||
GenBank mRNA | AF311338.1 | - | 5 | 634 | ||
AK027813.1 | - | 3 | 1924 | |||
AK127891.1 | - | 2 | 2015 | |||
AY358525.1 | - | 5 | 538 | |||
BC006292.2 | - | 2 | 251 | |||
BC070231.1 | - | 5 | 549 | 6617879 | ||
BC119730.1 | - | 2 | 271 | |||
BC119731.1 | - | 2 | 271 | |||
BC127649.1 | - | 5 | 537 | 40118154 | ||
BC127650.1 | - | 2 | 271 | |||
BM920977.1 | - | 5 | 531 | |||
BU852346.1 | - | 5 | 552 |