Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TMEM107 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_032354.3 | NP_115730.2 | 5 | 1214 | ||
NM_183065.2 | NP_898888.1 | 5 | 1196 | |||
GenBank mRNA | BC006292.2 | - | 2 | 887 |
Gene Symbol: SNORD118 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_033294.1 | 1 | 49 |