Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MEGF11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_032445.2 | NP_115821.2 | 16 | 2118 | ||
XM_017022670.1 | XP_016878159.1 | 15 | 2540 | |||
XM_017022671.1 | XP_016878160.1 | 15 | 2538 | |||
XM_017022672.1 | XP_016878161.1 | 15 | 2547 | |||
XM_017022673.1 | XP_016878162.1 | 15 | 2541 | |||
XM_017022674.1 | XP_016878163.1 | 10 | 2514 | |||
XM_017022675.1 | XP_016878164.1 | 10 | 2552 | |||
GenBank mRNA | AB058677.1 | - | 14 | 1911 | ||
AB300051.1 | - | 15 | 1977 | |||
AL834326.1 | - | 9 | 1232 | |||
BC029999.1 | - | 16 | 2150 | |||
BC117419.1 | - | 16 | 2038 | 40125970 | ||
BC126313.1 | - | 16 | 2038 | 8992029 |