Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GHDC | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001142623.1 | NP_001136095.1 | 4 | 517 | ||
NM_032484.4 | NP_115873.1 | 4 | 517 | |||
GenBank mRNA | AF316997.1 | - | 2 | 293 | ||
AK074700.1 | - | 3 | 721 | |||
AK075277.1 | - | 4 | 479 | |||
AK122777.1 | - | 5 | 285 | |||
AK123075.1 | - | 678 | ||||
AK298933.1 | - | 4 | 395 | |||
AK307058.1 | - | 4 | 537 | |||
AK307628.1 | - | 3 | 765 | |||
BC022784.1 | - | 4 | 452 | 3847323 |