Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SPATA22 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001170695.1 | NP_001164166.1 | 9 | 1112 | ||
NM_001170696.1 | NP_001164167.1 | 8 | 983 | |||
NM_001170697.1 | NP_001164168.1 | 9 | 1118 | |||
NM_001170698.1 | NP_001164169.1 | 9 | 1133 | |||
NM_001170699.1 | NP_001164170.1 | 8 | 1254 | |||
NM_001321336.1 | NP_001308265.1 | 9 | 1402 | |||
NM_001321337.1 | NP_001308266.1 | 9 | 1450 | |||
NM_032598.4 | NP_115987.2 | 9 | 1174 | |||
GenBank mRNA | AF367472.1 | - | 9 | 1066 | ||
AK057485.1 | - | 1 | 1541 | |||
AK295299.1 | - | 9 | 1402 | |||
AK301892.1 | - | 8 | 1220 | |||
AY032684.1 | - | 8 | 932 | |||
AY035867.1 | - | 9 | 1115 | |||
AY035868.1 | - | 9 | 1174 | |||
BC029483.1 | - | 9 | 1059 | 5271286 |