Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CRACR2A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001144958.1 | NP_001138430.1 | 7 | 1144 | ||
NM_032680.3 | NP_116069.1 | 7 | 1144 | |||
XM_006719021.3 | XP_006719084.1 | 7 | 1144 | |||
XM_011521034.2 | XP_011519336.1 | 6 | 891 | |||
XM_011521036.2 | XP_011519338.1 | 5 | 651 | |||
GenBank mRNA | AK304017.1 | - | 7 | 1046 | ||
BC004524.2 | - | 7 | 1041 | |||
BC150643.1 | - | 6 | 775 | 9057013 |