Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TMTC4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001079669.2 | NP_001073137.1 | 14 | 1996 | ||
NM_001286453.1 | NP_001273382.1 | 12 | 1663 | |||
NM_032813.3 | NP_116202.2 | 15 | 2206 | |||
XM_011521121.2 | XP_011519423.1 | 16 | 2358 | |||
XM_011521122.2 | XP_011519424.1 | 16 | 2274 | |||
XM_011521123.2 | XP_011519425.1 | 16 | 2435 | |||
XM_011521124.2 | XP_011519426.1 | 15 | 2063 | |||
XM_011521125.1 | XP_011519427.1 | 16 | 2267 | |||
XM_017020796.1 | XP_016876285.1 | 15 | 2261 | |||
XM_017020797.1 | XP_016876286.1 | 14 | 2000 | |||
XM_017020798.1 | XP_016876287.1 | 14 | 2096 | |||
XM_017020799.1 | XP_016876288.1 | 13 | 1781 | |||
GenBank mRNA | AF339792.1 | - | 12 | 1578 | ||
AK027530.1 | - | 9 | 1089 | |||
AK056409.1 | - | 15 | 2065 | |||
AK092653.1 | - | 7 | 2079 | |||
AK290345.1 | - | 14 | 1856 | |||
AK299859.1 | - | 12 | 1502 | |||
BC018707.1 | - | 9 | 1167 | 4841071 | ||
BX647956.1 | - | 14 | 1887 | |||
BX648008.1 | - | 14 | 1840 | |||
DA368450.1 | - | 2 | 160 |