Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FAM104A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001098832.1 | NP_001092302.1 | 4 | 1300 | ||
NM_001289410.1 | NP_001276339.1 | 2 | 1137 | |||
NM_001289411.1 | NP_001276340.1 | 3 | 1155 | |||
NM_001289412.1 | NP_001276341.1 | 4 | 1073 | |||
NM_032837.2 | NP_116226.2 | 3 | 1237 | |||
GenBank mRNA | AK027681.1 | - | 3 | 1122 | ||
AK074755.1 | - | 3 | 1123 | |||
BC011054.1 | - | 3 | 962 | 4177665 | ||
BC025238.1 | - | 3 | 1000 | 4905931 | ||
BX537858.1 | - | 4 | 1204 | |||
BX648968.1 | - | 5 | 1216 |