Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MEDAG | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_032849.3 | NP_116238.2 | 3 | 741 | ||
XM_017020801.1 | XP_016876290.1 | 2 | 1121 | |||
GenBank mRNA | AB055407.1 | - | 2 | 127 | ||
AK027740.1 | - | 340 | ||||
AK055635.1 | - | 3 | 725 | |||
BC040165.1 | - | 3 | 625 | 5590156 | ||
BC101624.1 | - | 3 | 479 | 8069130 | ||
BC101626.1 | - | 3 | 479 | 8069132 |