Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TNS4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_032865.5 | NP_116254.4 | 13 | 2908 | ||
XM_005257744.1 | XP_005257801.1 | 13 | 2984 | |||
XM_017025236.1 | XP_016880725.1 | 12 | 2909 | |||
GenBank mRNA | AF370421.1 | - | 13 | 2902 | ||
AF417488.1 | - | 13 | 2870 | |||
AK027856.1 | - | 11 | 2258 | |||
AW937802.1 | - | 1 | 125 | |||
BC013373.2 | - | 4 | 1020 | |||
BC018706.1 | - | 12 | 2354 | 5013235 |