Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DRG1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_004147.3 | NP_004138.1 | 2 | 205 | ||
GenBank mRNA | AF078103.1 | - | 2 | 135 | ||
AJ005940.1 | - | 2 | 139 | |||
AK315659.1 | - | 2 | 163 | |||
BC019285.1 | - | 2 | 144 | 2823263 | ||
BC020803.1 | - | 2 | 139 | 4250998 | ||
BT007237.1 | - | 2 | 74 | |||
CR456488.1 | - | 2 | 116 | |||
CR542059.1 | - | 2 | 74 |