Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZCRB1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_033114.3 | NP_149105.3 | 7, 8 | 724 | ||
XM_017020124.1 | XP_016875613.1 | 6, 7 | 615 | |||
GenBank mRNA | AB062361.1 | - | 7, 8 | 724 | ||
AK308244.1 | - | 595 | ||||
BC022543.1 | - | 7, 8 | 699 | 4797962 | ||
BI666919.1 | - | 3, 4 | 263 |