Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ATP23 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001320408.1 | NP_001307337.1 | 6 | 960 | ||
NM_001320409.1 | NP_001307338.1 | 5 | 874 | |||
NM_001320410.1 | NP_001307339.1 | 7 | 1091 | |||
NM_033276.3 | NP_150592.1 | 6 | 920 | |||
XM_011538976.2 | XP_011537278.1 | 5 | 595 | |||
XM_017020206.1 | XP_016875695.1 | 6 | 710 | |||
GenBank mRNA | AF078164.2 | - | 6 | 850 | ||
BC012776.2 | - | 6 | 762 | 3936722 | ||
BC115381.1 | - | 6 | 761 | 40036852 | ||
BC115382.1 | - | 6 | 760 | 40036859 |