Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: C15orf57 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001080791.2 | NP_001074260.1 | 3 | 639 | ||
NM_001080792.2 | NP_001074261.1 | 3 | 429 | |||
NM_001289132.1 | NP_001276061.1 | 3 | 614 | |||
NM_052849.3 | NP_443081.1 | 3 | 614 | |||
GenBank mRNA | AK092214.1 | - | 3 | 562 | ||
AK098781.1 | - | 3 | 549 | |||
AK293079.1 | - | 3 | 414 | |||
AK312842.1 | - | 3 | 404 | |||
AL832032.2 | - | 2 | 361 | |||
BC001673.2 | - | 2 | 353 | 2823236 | ||
BG709516.1 | - | 3 | 432 | |||
BP202606.1 | - | 3 | 406 | |||
BP289634.1 | - | 3 | 429 | |||
BQ878848.1 | - | 3 | 388 | |||
BU657334.1 | - | 3 | 403 | |||
BX404085.2 | - | 3 | 608 | |||
CD686743.1 | - | 1 | 74 |