Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other IFFO1 SIRNA Assays ›
Gene Symbol:
IFFO1
Gene Name:
intermediate filament family orphan 1
Gene Aliases:
HOM-TES-103, IFFO
Chromosome Location:
Chr.12: 6539528 - 6556102 on Build GRCh38
UniGene ID:
Hs.15243
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
141244
Purification
HPLC
Standard
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: IFFO1
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001039670.2
NP_001034759.1
10
2459
NM_001193457.1
NP_001180386.1
10
2483
NM_080730.4
NP_542768.2
10
2456
NR_036467.1
10
2556
XM_006718975.3
XP_006719038.1
10
2475
XM_006718976.3
XP_006719039.1
9
2469
XM_006718977.3
XP_006719040.1
9
2466
XM_006718981.3
XP_006719044.1
11
2144
XM_011520947.2
XP_011519249.1
9
2742
XM_011520948.2
XP_011519250.1
9
2739
XM_011520949.2
XP_011519251.1
8
2733
XM_011520952.2
XP_011519254.1
10
2408
XM_017019170.1
XP_016874659.1
9
2545
XM_017019171.1
XP_016874660.1
10
2472
XM_017019172.1
XP_016874661.1
10
2411
XM_017019173.1
XP_016874662.1
9
2042
GenBank mRNA
AF124432.1
-
10
2114
AK000710.1
-
5
1687
AK095106.1
-
6
2034
AL080214.1
-
3
1032
BC001790.1
-
10
2536
BC002857.2
-
9
1597
BC004384.1
-
9
1597
BC010431.1
-
10
2536
BC023664.2
-
10
2536
BC063004.1
-
10
1730
BC071170.1
-
5
1608
BC103817.1
-
5
1613
BC103818.1
-
5
1613
BC110387.1
-
10
2449
5183989
BC113848.1
-
5
1613
BC114454.1
-
5
1610
BX648701.1
-
11
2045
BX648743.1
-
11
2045
Back To Top
More Information
Panther Classification:
Molecular Function -
dehydrogenase
oxidoreductase
metabolite interconversion enzyme
Gene Ontology Categories:
Process(es)
structural molecule activity
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-blue-69b65fdbbd-sq4fc:80/100.66.76.194:80.
git-commit: a4c025c8ba47546866237b65de1f276a5fdbbe78
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.18.1-Offline