Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: WIPF2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_133264.4 | NP_573571.1 | 8 | 1661 | ||
XM_005257083.2 | XP_005257140.1 | 8 | 1663 | |||
XM_005257084.2 | XP_005257141.1 | 8 | 1621 | |||
XM_011524412.1 | XP_011522714.1 | 7 | 1562 | |||
XM_011524413.1 | XP_011522715.1 | 7 | 1543 | |||
GenBank mRNA | AF370396.1 | - | 8 | 1586 | ||
AK026913.1 | - | 6 | 1109 | |||
AK055057.1 | - | 6 | 878 | |||
AK289779.1 | - | 8 | 1680 | |||
AK290301.1 | - | 8 | 1657 | |||
AK315836.1 | - | 8 | 1641 | |||
AL834239.1 | - | 7 | 1530 | |||
BC025965.1 | - | 8 | 1617 | |||
BC065551.1 | - | 8 | 1598 | 6044222 | ||
U90911.1 | - | 1 | 84 |