Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DIS3L | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001143688.2 | NP_001137160.1 | 16, 17 | 2885 | ||
NM_001323936.1 | NP_001310865.1 | 16, 17 | 2804 | |||
NM_001323937.1 | NP_001310866.1 | 15, 16 | 2706 | |||
NM_001323938.1 | NP_001310867.1 | 16, 17 | 2812 | |||
NM_001323939.1 | NP_001310868.1 | 17, 18 | 3009 | |||
NM_001323940.1 | NP_001310869.1 | 15, 16 | 2715 | |||
NM_001323941.1 | NP_001310870.1 | 16, 17 | 3280 | |||
NM_001323943.1 | NP_001310872.1 | 15, 16 | 2909 | |||
NM_001323944.1 | NP_001310873.1 | 16, 17 | 3079 | |||
NM_001323945.1 | NP_001310874.1 | 16, 17 | 3353 | |||
NM_001323946.1 | NP_001310875.1 | 16, 17 | 3006 | |||
NM_001323948.1 | NP_001310877.1 | 15, 16 | 2950 | |||
NM_133375.4 | NP_588616.1 | 16, 17 | 2835 | |||
GenBank mRNA | AB075835.1 | - | 15, 16 | 2542 | ||
BC014124.2 | - | 8, 9 | 1633 | |||
BC022089.2 | - | 16, 17 | 2770 | 3029342 |