Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FAM181A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001207071.1 | NP_001194000.1 | 2 | 683 | ||
NM_001207072.1 | NP_001194001.1 | 2 | 824 | |||
NM_001207073.1 | NP_001194002.1 | 2 | 694 | |||
NM_001207074.1 | NP_001194003.1 | 2 | 640 | |||
NM_138344.4 | NP_612353.3 | 3 | 1006 | |||
XM_011537297.1 | XP_011535599.1 | 2 | 657 | |||
GenBank mRNA | AK093387.1 | - | 3 | 991 | ||
AK315393.1 | - | 2 | 694 | |||
AY255795.1 | - | 2 | 660 | |||
BC009073.1 | - | 2 | 824 | 3856004 | ||
CX870424.1 | - | 2 | 640 |