Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CASC4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_138423.3 | NP_612432.2 | 7 | 1136 | ||
NM_177974.2 | NP_816929.1 | 7 | 1136 | |||
GenBank mRNA | AK095068.1 | - | 3 | 176 | ||
AK291026.1 | - | 7 | 893 | |||
AK298564.1 | - | 8 | 851 | |||
AL832333.1 | - | 7 | 908 | |||
AL832840.1 | - | 7 | 578 | |||
AY358086.1 | - | 7 | 1087 | |||
BC041786.1 | - | 14 | ||||
BC063480.1 | - | 7 | 1090 | 5218924 |