Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZNF689 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_138447.2 | NP_612456.1 | 2 | 600 | ||
NR_073480.1 | 2 | 235 | ||||
NR_073481.1 | 2 | 238 | ||||
NR_073482.1 | 2 | 118 | ||||
NR_104166.1 | 2 | 137 | ||||
GenBank mRNA | AB065282.1 | - | 2 | 588 | ||
AK074896.1 | - | 2 | 323 | |||
AK310459.1 | - | 2 | 157 | |||
BC014000.2 | - | 2 | 575 | 3936339 | ||
CN406120.1 | - | 2 | 249 | |||
DA010230.1 | - | 2 | 239 | |||
DA485303.1 | - | 2 | 235 | |||
DB456506.1 | - | 2 | 118 | |||
HY005770.1 | - | 2 | 132 |