Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SPPL3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_139015.4 | NP_620584.2 | 3, 4 | 675 | ||
XM_011537925.2 | XP_011536227.1 | 3, 4 | 205 | |||
GenBank mRNA | AB252457.1 | - | 3, 4 | 267 | ||
AJ345030.1 | - | 3, 4 | 243 | |||
AK074916.1 | - | 2, 3 | 158 | |||
AK223590.1 | - | 3, 4 | 671 | |||
AK291161.1 | - | 3, 4 | 497 | |||
AY169313.1 | - | 3, 4 | 449 | |||
BC073910.1 | - | 3, 4 | 456 | 4538522 | ||
BC101625.1 | - | 3, 4 | 270 | 8069131 | ||
BC101627.1 | - | 3, 4 | 270 | 8069133 | ||
BU932077.1 | - | 3, 4 | 675 |