Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LRRC28 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001284400.2 | NP_001271329.1 | 3, 4 | 446 | ||
NM_001321675.1 | NP_001308604.1 | 3, 4 | 442 | |||
NM_001321676.1 | NP_001308605.1 | 3, 4 | 446 | |||
NM_001321677.1 | NP_001308606.1 | 3, 4 | 446 | |||
NM_001321678.1 | NP_001308607.1 | 3, 4 | 294 | |||
NM_144598.4 | NP_653199.2 | 3, 4 | 446 | |||
NR_135753.1 | 3, 4 | 446 | ||||
NR_135754.1 | 3, 4 | 294 | ||||
NR_135757.1 | 3, 4 | 446 | ||||
XM_006720389.3 | XP_006720452.1 | 3, 4 | 446 | |||
XM_011521218.2 | XP_011519520.1 | 3, 4 | 482 | |||
XM_011521220.1 | XP_011519522.1 | 3, 4 | 446 | |||
XM_011521221.2 | XP_011519523.1 | 3, 4 | 446 | |||
XM_017021914.1 | XP_016877403.1 | 3, 4 | 730 | |||
GenBank mRNA | AK292887.1 | - | 3, 4 | 303 | ||
AK293832.1 | - | 3, 4 | 303 | |||
AK295161.1 | - | 3, 4 | 321 | |||
AK296352.1 | - | 3, 4 | 348 | |||
AY358082.1 | - | 3, 4 | 303 | |||
BC047306.1 | - | 3, 4 | 361 | 4999965 |