Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZC3H18 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001294340.1 | NP_001281269.1 | 8 | 1468 | ||
NM_144604.3 | NP_653205.3 | 7 | 1396 | |||
XM_011522865.1 | XP_011521167.1 | 1 | 107 | |||
XM_017022934.1 | XP_016878423.1 | 8 | 1468 | |||
GenBank mRNA | AK091849.1 | - | 2 | 223 | ||
AK122593.1 | - | 1 | 525 | |||
AK225807.1 | - | 7 | 1366 | |||
AK300254.1 | - | 8 | 1446 | |||
AK302716.1 | - | 7 | 1023 | |||
BC001584.1 | - | 4 | 492 | |||
BC050463.1 | - | 7 | 1343 | 6042036 | ||
DB007291.1 | - | 4 | 482 |